Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284413.1 | NP_001271342.1 | 4 | 661 | ||
NM_001284415.1 | NP_001271344.1 | 5 | 742 | |||
NM_012260.3 | NP_036392.2 | 5 | 742 | |||
GenBank mRNA | AF161397.1 | - | 5 | 362 | ||
AJ131753.1 | - | 5 | 452 | |||
AK301546.1 | - | 4 | 378 | |||
AK302086.1 | - | 5 | 459 | |||
BC001627.1 | - | 5 | 460 | 3538159 | ||
DA609383.1 | - | 5 | 459 | |||
DB049608.1 | - | 5 | 513 |