Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012455.2 | NP_036587.2 | 17 | 4206 | ||
XM_005263634.2 | XP_005263691.1 | 17 | 4271 | |||
XM_006712392.2 | XP_006712455.1 | 17 | 4215 | |||
XM_006712393.2 | XP_006712456.1 | 16 | 4073 | |||
XM_006712394.2 | XP_006712457.1 | 16 | 4070 | |||
GenBank mRNA | AB208798.1 | - | 16 | 4642 | ||
AK093556.1 | - | 2 | 1779 | |||
AK094598.1 | - | 4 | 2118 | |||
BC035307.1 | - | 17 | 4105 | 4339380 | ||
BC073151.1 | - | 17 | 4168 | 5757431 | ||
U63127.1 | - | 16 | 4090 |