Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 6 | 1117 | ||
NM_001271780.1 | NP_001258709.1 | 5 | 832 | |||
NM_001271781.1 | NP_001258710.1 | 5 | 707 | |||
NM_014814.2 | NP_055629.1 | 5 | 958 | |||
XM_005265619.1 | XP_005265676.1 | 5 | 1054 | |||
XM_011534288.1 | XP_011532590.1 | 6 | 1013 | |||
XM_017007569.1 | XP_016863058.1 | 6 | 860 | |||
GenBank mRNA | AF215935.1 | - | 5 | 824 | ||
AF530062.1 | - | 5 | 771 | |||
AK095059.1 | - | 4 | 1540 | |||
AK290205.1 | - | 5 | 812 | |||
AL529900.3 | - | 5 | 693 | |||
AY359879.1 | - | 5 | 804 | |||
AY568086.1 | - | 5 | 796 | |||
BC000630.1 | - | 5 | 788 | 3344973 | ||
BC000904.2 | - | 5 | 809 | 2900971 | ||
BC012369.1 | - | 5 | 806 | 3858094 | ||
BX647742.1 | - | 6 | 984 | |||
D14663.1 | - | 5 | 796 |