Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 9 | 1526 | ||
NM_001271780.1 | NP_001258709.1 | 8 | 1241 | |||
NM_001271781.1 | NP_001258710.1 | 8 | 1116 | |||
NM_014814.2 | NP_055629.1 | 8 | 1367 | |||
XM_005265619.1 | XP_005265676.1 | 8 | 1463 | |||
XM_011534288.1 | XP_011532590.1 | 8 | 1253 | |||
XM_017007569.1 | XP_016863058.1 | 9 | 1269 | |||
GenBank mRNA | AF215935.1 | - | 8 | 1233 | ||
AK054730.1 | - | 2 | 2518 | |||
AK055210.1 | - | 4 | 1869 | |||
AK095059.1 | - | 7 | 1949 | |||
AK290205.1 | - | 8 | 1221 | |||
AL832109.1 | - | 3 | 4477 | |||
AY359879.1 | - | 8 | 1213 | |||
AY568086.1 | - | 8 | 1205 | |||
BC000630.1 | - | 8 | 1197 | 3344973 | ||
BC000904.2 | - | 8 | 1218 | 2900971 | ||
BC012369.1 | - | 8 | 1215 | 3858094 | ||
BX647742.1 | - | 9 | 1393 | |||
CD110455.1 | - | 2 | 159 | |||
CD678025.1 | - | 1 | 432 | |||
D14663.1 | - | 8 | 1205 |