Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161580.1 | NP_001155052.1 | 10 | 2009 | ||
NM_001161581.1 | NP_001155053.1 | 11 | 2224 | |||
NM_015426.4 | NP_056241.3 | 11 | 2153 | |||
XM_011533560.1 | XP_011531862.1 | 10 | 1776 | |||
XM_011533561.1 | XP_011531863.1 | 9 | 1632 | |||
XM_017006104.1 | XP_016861593.1 | 10 | 1802 | |||
GenBank mRNA | AF088072.1 | - | 1 | 614 | ||
AK075289.1 | - | 11 | 1868 | |||
BC110877.1 | - | 10 | 2009 | 5588109 |