Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI3BP SIRNA Assays ›
Gene Symbol:
ABI3BP
Gene Name:
ABI family member 3 binding protein
Gene Aliases:
NESHBP, TARSH
Chromosome Location:
Chr.3: 100749335 - 100993508 on Build GRCh38
UniGene ID:
Hs.477015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124875
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI3BP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015429.3
NP_056244.2
2
308
XM_005247281.2
XP_005247338.1
2
311
XM_005247282.2
XP_005247339.1
2
311
XM_005247283.2
XP_005247340.1
2
311
XM_005247284.2
XP_005247341.1
2
311
XM_005247285.2
XP_005247342.1
2
311
XM_005247287.2
XP_005247344.1
2
311
XM_005247288.2
XP_005247345.1
2
311
XM_005247289.2
XP_005247346.1
2
311
XM_005247290.2
XP_005247347.1
2
311
XM_005247291.2
XP_005247348.1
2
311
XM_005247293.2
XP_005247350.1
2
311
XM_005247294.2
XP_005247351.1
2
311
XM_005247295.2
XP_005247352.1
2
311
XM_005247296.2
XP_005247353.1
2
311
XM_005247298.2
XP_005247355.1
2
311
XM_005247299.3
XP_005247356.1
2
326
XM_005247300.2
XP_005247357.1
2
311
XM_005247301.2
XP_005247358.1
2
311
XM_005247302.2
XP_005247359.1
2
311
XM_005247303.2
XP_005247360.1
2
311
XM_005247305.2
XP_005247362.1
2
311
XM_005247306.2
XP_005247363.1
2
311
XM_006713569.2
XP_006713632.1
2
311
XM_006713570.2
XP_006713633.1
2
311
XM_006713571.2
XP_006713634.1
2
311
XM_006713572.2
XP_006713635.1
2
311
XM_011512639.1
XP_011510941.1
2
311
XM_011512640.1
XP_011510942.1
2
311
XM_011512641.1
XP_011510943.1
2
311
XM_011512643.1
XP_011510945.1
2
311
XM_011512644.1
XP_011510946.1
2
311
XM_011512645.1
XP_011510947.1
2
311
XM_011512646.1
XP_011510948.1
2
311
XM_011512647.1
XP_011510949.1
2
311
XM_011512648.1
XP_011510950.1
2
311
XM_011512649.1
XP_011510951.1
2
311
XM_011512651.1
XP_011510953.1
2
311
XM_011512652.1
XP_011510954.1
2
311
XM_011512653.1
XP_011510955.1
2
311
XM_011512654.1
XP_011510956.1
2
311
XM_011512656.1
XP_011510958.1
2
311
XM_011512657.1
XP_011510959.1
2
311
XM_011512658.1
XP_011510960.1
2
311
XM_011512659.1
XP_011510961.1
2
311
XM_011512660.1
XP_011510962.1
2
311
XM_017006105.1
XP_016861594.1
2
311
XM_017006106.1
XP_016861595.1
2
311
XM_017006107.1
XP_016861596.1
2
311
XM_017006108.1
XP_016861597.1
2
311
GenBank mRNA
AK025204.1
-
2
268
AL833204.1
-
2
271
BC030221.1
-
2
288
5205716
BX648726.1
-
1
15
DA342832.1
-
2
308
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline