Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199570.1 | NP_001186499.1 | 5, 6 | 1002 | ||
NM_015526.2 | NP_056341.1 | 6, 7 | 918 | |||
GenBank mRNA | AJ427922.1 | - | 6, 7 | 896 | ||
AK056445.1 | - | 4, 5 | 402 | |||
AK090732.1 | - | 6, 7 | 921 | |||
AK094738.1 | - | 6, 7 | 916 | |||
AK289509.1 | - | 5, 6 | 970 | |||
AK294748.1 | - | 6, 7 | 789 | |||
BC013116.1 | - | 6, 7 | 830 | 4214441 | ||
BC014486.1 | - | 3, 4 | 361 | |||
BC128090.1 | - | 2, 3 | 170 |