Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIBC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015653.4 | NP_056468.3 | 7 | 1384 | ||
XM_005261524.4 | XP_005261581.1 | 7 | 1209 | |||
XM_011530126.2 | XP_011528428.1 | 5 | 912 | |||
GenBank mRNA | AK098586.1 | - | 7 | 1384 | ||
AK225722.1 | - | 7 | 1365 | |||
AL050075.1 | - | 3 | 492 | |||
AL442116.1 | - | 7 | 1335 | |||
BC003024.1 | - | 7 | 1346 | 2822570 | ||
BC009904.2 | - | 7 | 1346 | 2822570 |