Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127397.2 | NP_001120869.1 | 7 | 835 | ||
NM_001127398.2 | NP_001120870.1 | 7 | 835 | |||
NM_015701.4 | NP_056516.2 | 7 | 835 | |||
XM_011532766.1 | XP_011531068.1 | 7 | 749 | |||
GenBank mRNA | AF131849.1 | - | 1 | 24 | ||
AK001913.1 | - | 7 | 716 | |||
AK315477.1 | - | 7 | 624 | |||
AY358717.1 | - | 7 | 719 | |||
AY453410.1 | - | 7 | 686 | |||
BC013129.1 | - | 7 | 686 | 3447840 | ||
BC022228.1 | - | 7 | 683 | 4447432 | ||
EU794676.1 | - | 7 | 835 |