Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127397.2 | NP_001120869.1 | 11 | 1443 | ||
NM_001127398.2 | NP_001120870.1 | 10 | 1281 | |||
NM_015701.4 | NP_056516.2 | 11 | 1443 | |||
XM_011532766.1 | XP_011531068.1 | 11 | 1357 | |||
GenBank mRNA | AF131743.1 | - | 3 | 262 | ||
AF131849.1 | - | 5 | 632 | |||
AK001913.1 | - | 11 | 1324 | |||
AK315477.1 | - | 11 | 1232 | |||
AY358717.1 | - | 10 | 1165 | |||
AY453410.1 | - | 11 | 1294 | |||
BC013129.1 | - | 11 | 1294 | 3447840 | ||
BC022228.1 | - | 10 | 1129 | 4447432 | ||
EU794676.1 | - | 11 | 1443 |