Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VILL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015873.3 | NP_056957.3 | 12 | 1465 | ||
XM_005265191.3 | XP_005265248.1 | 13 | 1573 | |||
XM_005265192.4 | XP_005265249.1 | 13 | 2708 | |||
XM_006713184.3 | XP_006713247.1 | 14 | 1573 | |||
XM_011533770.2 | XP_011532072.1 | 13 | 1729 | |||
XM_011533771.1 | XP_011532073.1 | 13 | 1729 | |||
XM_011533773.2 | XP_011532075.1 | 13 | 1730 | |||
GenBank mRNA | AB209574.1 | - | 1 | 854 | ||
AK309304.1 | - | 10 | 1355 | |||
BC000243.1 | - | 9 | 1001 | 3356294 | ||
BC004300.1 | - | 13 | 1495 | 3622951 | ||
D88154.1 | - | 8 | 871 | |||
KU178619.1 | - | 8 | 869 | |||
KU178620.1 | - | 8 | 869 |