Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015934.4 | NP_057018.1 | 4 | 418 | ||
GenBank mRNA | AF123534.1 | - | 4 | 192 | ||
AF263608.1 | - | 4 | 192 | |||
AK023975.1 | - | 3 | 117 | |||
AK300844.1 | - | 4 | 398 | |||
AK307396.1 | - | 4 | 398 | |||
AL117554.1 | - | 4 | 401 | |||
BC001707.1 | - | 4 | 384 | |||
BC009306.2 | - | 4 | 348 | |||
BC018805.1 | - | 4 | 371 | |||
BC032592.2 | - | 4 | 342 | 5527148 | ||
BP245255.1 | - | 4 | 418 |