Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015934.4 | NP_057018.1 | 5, 6 | 668 | ||
GenBank mRNA | AF123534.1 | - | 5, 6 | 442 | ||
AF161469.1 | - | 3, 4 | 324 | |||
AF263608.1 | - | 5, 6 | 442 | |||
AK023975.1 | - | 4, 5 | 367 | |||
AK307396.1 | - | 5, 6 | 648 | |||
AL117554.1 | - | 5, 6 | 651 | |||
BC001707.1 | - | 5, 6 | 634 | |||
BC009306.2 | - | 5, 6 | 598 | |||
BC018805.1 | - | 5, 6 | 621 | |||
BC032592.2 | - | 5, 6 | 592 | 5527148 |