Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137602.2 | NP_001131074.1 | 3 | 662 | ||
NM_001301833.1 | NP_001288762.1 | 5 | 442 | |||
NM_001301852.1 | NP_001288781.1 | 4 | 364 | |||
NM_015955.3 | NP_057039.1 | 4 | 731 | |||
NR_126032.1 | 4 | 731 | ||||
NR_126034.1 | 4 | 364 | ||||
XM_011532892.2 | XP_011531194.1 | 5 | 379 | |||
XM_011532896.2 | XP_011531198.1 | 3 | 257 | |||
XM_017004255.1 | XP_016859744.1 | 5 | 352 | |||
GenBank mRNA | AB041018.1 | - | 4 | 731 | ||
AB041019.1 | - | 4 | 380 | |||
AF132961.1 | - | 4 | 429 | |||
AF363446.1 | - | 4 | 648 | |||
AF529368.1 | - | 4 | 314 | |||
AK057760.1 | - | 4 | 353 | |||
AK290753.1 | - | 5 | 398 | |||
AK297128.1 | - | 3 | 373 | |||
BC018733.1 | - | 4 | 696 | 4851517 | ||
BC070046.1 | - | 5 | 471 | 5273070 | ||
BC094681.1 | - | 4 | 335 | 6205399 | ||
BG033287.1 | - | 4 | 313 | |||
BP248979.1 | - | 5 | 442 | |||
CD250945.1 | - | 4 | 428 |