Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 | 579 | ||
NM_001127400.1 | NP_001120872.1 | 4 | 815 | |||
NM_001127401.1 | NP_001120873.1 | 5 | 876 | |||
NM_016061.2 | NP_057145.1 | 3 | 518 | |||
XM_017004318.1 | XP_016859807.1 | 5 | 1114 | |||
XM_017004319.1 | XP_016859808.1 | 4 | 552 | |||
XM_017004320.1 | XP_016859809.1 | 4 | 1053 | |||
XM_017004321.1 | XP_016859810.1 | 4 | 745 | |||
XM_017004322.1 | XP_016859811.1 | 4 | 976 | |||
GenBank mRNA | AB098739.1 | - | 2 | 315 | ||
AF135161.1 | - | 3 | 422 | |||
AF151885.1 | - | 3 | 440 | |||
AK025730.1 | - | 4 | 456 | |||
AK307099.1 | - | 2 | 231 | |||
AK311404.1 | - | 3 | 229 | |||
BC000836.1 | - | 4 | 393 | 3456031 | ||
BC047237.1 | - | 4 | 976 | 5269563 |