Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317747.1 | NP_001304676.1 | 5 | 541 | ||
NM_016388.3 | NP_057472.2 | 6 | 652 | |||
GenBank mRNA | AF161547.1 | - | 6 | 567 | ||
AJ224878.1 | - | 6 | 551 | |||
AJ240085.1 | - | 5 | 467 | |||
AK291991.1 | - | 6 | 652 | |||
BC025713.1 | - | 6 | 558 | 5227396 | ||
BX415345.2 | - | 6 | 560 |