Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016474.4 | NP_057558.3 | 9 | 966 | ||
NR_135523.1 | 8 | 871 | ||||
GenBank mRNA | AK091158.1 | - | 4 | 350 | ||
BC013999.2 | - | 9 | 942 | 3936914 | ||
BC033897.1 | - | 9 | 953 | 4812977 |