Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271838.1 | NP_001258767.1 | 4 | 516 | ||
NM_016625.3 | NP_057709.2 | 4 | 504 | |||
GenBank mRNA | AF208853.1 | - | 4 | 462 | ||
AK290334.1 | - | 4 | 484 | |||
BC006982.1 | - | 4 | 453 | 3997840 | ||
DA273191.1 | - | 4 | 506 |