Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271834.1 | NP_001258763.1 | 4 | 496 | ||
NM_001271838.1 | NP_001258767.1 | 5 | 696 | |||
NM_016625.3 | NP_057709.2 | 5 | 684 | |||
GenBank mRNA | AK290334.1 | - | 5 | 664 | ||
AK300388.1 | - | 4 | 406 | |||
BC006982.1 | - | 5 | 633 | 3997840 | ||
BC010357.1 | - | 4 | 445 | 4249317 |