Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134422.1 | NP_001127894.1 | 5 | 1887 | ||
NM_001134423.2 | NP_001127895.1 | 5 | 1348 | |||
NM_001282762.1 | NP_001269691.1 | 5 | 1046 | |||
NM_001282763.1 | NP_001269692.1 | 5 | 573 | |||
NM_001282765.1 | NP_001269694.1 | 5 | 1506 | |||
NM_017548.4 | NP_060018.1 | 5 | 1150 | |||
XM_005247588.4 | XP_005247645.1 | 6 | 1180 | |||
XM_005247589.4 | XP_005247646.1 | 6 | 1177 | |||
XM_005247590.4 | XP_005247647.1 | 5 | 1174 | |||
XM_005247592.4 | XP_005247649.1 | 6 | 654 | |||
XM_005247594.4 | XP_005247651.1 | 6 | 558 | |||
XM_011512961.2 | XP_011511263.1 | 5 | 1911 | |||
XM_011512962.2 | XP_011511264.1 | 6 | 482 | |||
XM_017006808.1 | XP_016862297.1 | 5 | 1342 | |||
GenBank mRNA | AF103803.1 | - | 5 | 1018 | ||
AK025647.1 | - | 2 | 169 | |||
AK096865.1 | - | 5 | 1326 | |||
BG773216.1 | - | 5 | 578 | |||
CB268158.1 | - | 1 | 428 |