Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017590.5 | NP_060060.3 | 4 | 533 | ||
XM_011530016.2 | XP_011528318.1 | 4 | 514 | |||
GenBank mRNA | AB028954.1 | - | 1 | 146 | ||
AK023849.1 | - | 4 | 321 | |||
AK225891.1 | - | 4 | 506 | |||
AK315009.1 | - | 4 | 475 | |||
BC098378.1 | - | 4 | 471 | |||
BC152558.1 | - | 4 | 511 | 8860405 |