Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017694.3 | NP_060164.3 | 8 | 3206 | ||
XM_005246654.2 | XP_005246711.1 | 7 | 3084 | |||
XM_005246656.3 | XP_005246713.1 | 8 | 3109 | |||
XM_011511368.1 | XP_011509670.1 | 9 | 3272 | |||
XM_017004392.1 | XP_016859881.1 | 9 | 3231 | |||
GenBank mRNA | AK000167.1 | - | 6 | 1984 | ||
AK021420.1 | - | 4 | 674 | |||
AK098085.1 | - | 6 | 2616 | |||
AK127176.1 | - | 8 | 3206 | |||
BC050537.2 | - | 6 | 2145 | 6204874 |