Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083590.1 | NP_001077059.1 | 11 | 1342 | ||
NM_017727.4 | NP_060197.4 | 12 | 1477 | |||
XM_005264381.3 | XP_005264438.1 | 13 | 1642 | |||
XM_005264382.3 | XP_005264439.1 | 13 | 1528 | |||
XM_005264383.3 | XP_005264440.1 | 5 | 525 | |||
GenBank mRNA | AF289555.1 | - | 6 | 513 | ||
AK000261.1 | - | 7 | 594 | |||
AK074981.1 | - | 12 | 1405 | |||
AK097308.1 | - | 11 | 1272 | |||
AK296575.1 | - | 5 | 577 | |||
AK297150.1 | - | 6 | 729 | |||
AK315359.1 | - | 10 | 941 | |||
AL833725.1 | - | 12 | 1337 | |||
AL834157.1 | - | 7 | 582 | |||
BC068479.1 | - | 12 | 1429 | 4814193 |