Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003093.2 | NP_003084.1 | 3 | 541 | ||
NR_029472.1 | 2 | 498 | ||||
GenBank mRNA | AK311955.1 | - | 3 | 121 | ||
BC121082.1 | - | 3 | 110 | 40119776 | ||
BC121083.1 | - | 3 | 110 | 40119777 | ||
BU153661.1 | - | 2 | 106 | |||
BU508721.1 | - | 2 | 67 | |||
CD109745.1 | - | 3 | 558 | |||
M18465.1 | - | 1 | 19 | |||
X12517.1 | - | 3 | 106 |