Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017875.2 | NP_060345.2 | 4 | 836 | ||
XM_006713214.1 | XP_006713277.1 | 5 | 799 | |||
XM_011533869.2 | XP_011532171.1 | 5 | 1699 | |||
XM_017006710.1 | XP_016862199.1 | 4 | 835 | |||
GenBank mRNA | AK000558.1 | - | 4 | 812 | ||
AK225388.1 | - | 4 | 788 | |||
BC013194.1 | - | 4 | 497 | 3918744 | ||
CR457242.1 | - | 4 | 435 |