Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM118A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104595.1 | NP_001098065.1 | 10 | 1764 | ||
NM_017911.2 | NP_060381.2 | 9 | 1437 | |||
XM_011530247.2 | XP_011528549.1 | 9 | 1555 | |||
XM_011530254.2 | XP_011528556.1 | 9 | 1345 | |||
XM_017028841.1 | XP_016884330.1 | 9 | 1340 | |||
XM_017028842.1 | XP_016884331.1 | 8 | 1107 | |||
XM_017028843.1 | XP_016884332.1 | 8 | 1104 | |||
XM_017028844.1 | XP_016884333.1 | 7 | 1074 | |||
XM_017028845.1 | XP_016884334.1 | 10 | 1540 | |||
XM_017028846.1 | XP_016884335.1 | 9 | 1424 | |||
XM_017028847.1 | XP_016884336.1 | 9 | 2068 | |||
XM_017028848.1 | XP_016884337.1 | 9 | 1466 | |||
XM_017028849.1 | XP_016884338.1 | 9 | 1302 | |||
XM_017028850.1 | XP_016884339.1 | 9 | 1425 | |||
XM_017028851.1 | XP_016884340.1 | 9 | 2056 | |||
XM_017028852.1 | XP_016884341.1 | 9 | 1462 | |||
XM_017028854.1 | XP_016884343.1 | 8 | 1189 | |||
XM_017028861.1 | XP_016884350.1 | 9 | 1739 | |||
GenBank mRNA | AK000642.1 | - | 9 | 1407 | ||
AK125027.1 | - | 10 | 1982 | |||
AK302205.1 | - | 6 | 916 | |||
BC013696.1 | - | 9 | 1194 | 3860694 |