Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017916.2 | NP_060386.1 | 8, 9 | 1123 | ||
XM_006723254.2 | XP_006723317.1 | 6, 7 | 687 | |||
GenBank mRNA | AK000650.1 | - | 8, 9 | 1075 | ||
BC001108.2 | - | 8, 9 | 1053 | 2966990 |