Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017931.2 | NP_060401.2 | 5 | 463 | ||
XM_011530259.1 | XP_011528561.1 | 6 | 591 | |||
XM_011530260.2 | XP_011528562.1 | 5 | 463 | |||
GenBank mRNA | AK000706.1 | - | 5 | 419 | ||
BC018918.2 | - | 5 | 404 | 4130906 | ||
DA762669.1 | - | 5 | 465 |