Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 3, 4 | 404 | ||
GenBank mRNA | AK312162.1 | - | 3, 4 | 387 | ||
BC002503.2 | - | 3, 4 | 365 | 3051095 | ||
BC008424.1 | - | 3, 4 | 389 | 4134501 | ||
BT006825.1 | - | 3, 4 | 210 | |||
CD516700.1 | - | 3, 4 | 410 | |||
CN271682.1 | - | 3, 4 | 404 | |||
CR450294.1 | - | 3, 4 | 210 | |||
M55580.1 | - | 215 | ||||
M77693.1 | - | 3, 4 | 375 |