Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204831.1 | NP_001191760.1 | 10 | 1827 | ||
NM_001204832.1 | NP_001191761.1 | 9 | 1629 | |||
NM_001204833.1 | NP_001191762.1 | 8 | 1494 | |||
NM_001204834.1 | NP_001191763.1 | 9 | 1257 | |||
NM_018075.3 | NP_060545.3 | 10 | 1827 | |||
XM_011533885.2 | XP_011532187.2 | 10 | 1793 | |||
XM_011533887.2 | XP_011532189.1 | 10 | 1793 | |||
XM_011533889.2 | XP_011532191.1 | 10 | 1793 | |||
XM_011533890.2 | XP_011532192.1 | 10 | 1793 | |||
XM_017006716.1 | XP_016862205.1 | 10 | 1793 | |||
XM_017006717.1 | XP_016862206.1 | 10 | 1745 | |||
XM_017006718.1 | XP_016862207.1 | 10 | 1772 | |||
XM_017006719.1 | XP_016862208.1 | 9 | 1547 | |||
XM_017006720.1 | XP_016862209.1 | 10 | 1772 | |||
XM_017006721.1 | XP_016862210.1 | 9 | 1556 | |||
XM_017006722.1 | XP_016862211.1 | 9 | 1529 | |||
GenBank mRNA | AK096302.1 | - | 10 | 1827 | ||
AK131223.1 | - | 9 | 1545 | |||
AK292368.1 | - | 10 | 1712 | |||
AK295969.1 | - | 8 | 1429 | |||
AK297949.1 | - | 10 | 1770 | |||
AL832508.1 | - | 10 | 1694 | |||
BC038855.1 | - | 9 | 1155 |