Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125130
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
30
4848
XM_005265279.4
XP_005265336.1
33
5442
XM_005265280.3
XP_005265337.1
32
5259
XM_005265282.3
XP_005265339.1
33
5401
XM_005265283.3
XP_005265340.1
32
5256
XM_011533900.2
XP_011532202.1
35
5608
XM_011533902.2
XP_011532204.1
34
5540
XM_011533903.2
XP_011532205.1
34
5545
XM_017006725.1
XP_016862214.1
34
5515
XM_017006726.1
XP_016862215.1
35
5625
XM_017006727.1
XP_016862216.1
35
5570
XM_017006728.1
XP_016862217.1
34
5566
XM_017006729.1
XP_016862218.1
34
5971
XM_017006730.1
XP_016862219.1
35
5561
XM_017006731.1
XP_016862220.1
34
5544
XM_017006732.1
XP_016862221.1
34
5537
XM_017006733.1
XP_016862222.1
34
5934
XM_017006734.1
XP_016862223.1
34
5507
XM_017006735.1
XP_016862224.1
33
5928
XM_017006736.1
XP_016862225.1
33
5926
XM_017006737.1
XP_016862226.1
33
5848
XM_017006738.1
XP_016862227.1
33
5469
XM_017006739.1
XP_016862228.1
33
5845
XM_017006740.1
XP_016862229.1
33
5386
XM_017006741.1
XP_016862230.1
33
5477
XM_017006742.1
XP_016862231.1
33
5500
XM_017006743.1
XP_016862232.1
34
5899
XM_017006744.1
XP_016862233.1
33
5466
XM_017006745.1
XP_016862234.1
33
5820
XM_017006746.1
XP_016862235.1
33
5817
XM_017006747.1
XP_016862236.1
33
5879
XM_017006748.1
XP_016862237.1
32
5775
XM_017006749.1
XP_016862238.1
32
5341
XM_017006750.1
XP_016862239.1
31
5229
XM_017006751.1
XP_016862240.1
33
5402
XM_017006752.1
XP_016862241.1
33
5818
XM_017006753.1
XP_016862242.1
33
5808
XM_017006754.1
XP_016862243.1
33
5384
XM_017006755.1
XP_016862244.1
32
5312
XM_017006756.1
XP_016862245.1
32
5309
XM_017006757.1
XP_016862246.1
30
5035
XM_017006758.1
XP_016862247.1
30
5166
XM_017006759.1
XP_016862248.1
32
5237
XM_017006760.1
XP_016862249.1
32
5219
XM_017006761.1
XP_016862250.1
32
5228
XM_017006762.1
XP_016862251.1
32
5216
XM_017006763.1
XP_016862252.1
33
5278
XM_017006764.1
XP_016862253.1
31
5156
XM_017006765.1
XP_016862254.1
30
5035
XM_017006766.1
XP_016862255.1
31
5088
GenBank mRNA
AF177387.1
-
29
4893
AF197569.1
-
30
4830
AF225870.1
-
28
4797
AF225871.1
-
28
4678
AF225872.1
-
28
4737
AK001507.1
-
6
920
AY281068.1
-
30
5057
BC015323.1
-
7
1184
BC089409.1
-
8
1265
BC115010.1
-
29
4834
BC115011.1
-
29
4909
7962010
BC129934.1
-
30
4947
40082629
BC129935.1
-
29
4782
40082630
DA039085.1
-
1
66
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline