Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306147.1 | NP_001293076.1 | 5 | 585 | ||
NM_018243.3 | NP_060713.1 | 3 | 365 | |||
XM_006714259.3 | XP_006714322.1 | 3 | 231 | |||
XM_017008400.1 | XP_016863889.1 | 4 | 374 | |||
XM_017008401.1 | XP_016863890.1 | 4 | 386 | |||
XM_017008402.1 | XP_016863891.1 | 4 | 386 | |||
XM_017008403.1 | XP_016863892.1 | 3 | 326 | |||
XM_017008404.1 | XP_016863893.1 | 3 | 326 | |||
GenBank mRNA | AK001711.1 | - | 3 | 268 | ||
AK302700.1 | - | 5 | 462 | |||
AL110300.1 | - | 3 | 247 | |||
BC008083.2 | - | 3 | 351 | 2966709 | ||
BC063615.1 | - | 3 | 216 | 3848367 | ||
CR997215.1 | - | 3 | 365 | |||
DA413789.1 | - | 3 | 239 | |||
DA497699.1 | - | 3 | 263 | |||
GU727629.1 | - | 3 | 177 |