Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018266.2 | NP_060736.1 | 7 | 1389 | ||
NR_073506.1 | 8 | 1498 | ||||
XM_005247578.1 | XP_005247635.1 | 7 | 1354 | |||
XM_006713687.1 | XP_006713750.1 | 7 | 1297 | |||
XM_011512958.2 | XP_011511260.1 | 7 | 2025 | |||
XM_017006788.1 | XP_016862277.1 | 6 | 1270 | |||
GenBank mRNA | AK001764.1 | - | 7 | 1331 | ||
AK222950.1 | - | 7 | 1321 | |||
AK223248.1 | - | 7 | 1321 | |||
BC003192.1 | - | 8 | 1413 | |||
BC010070.2 | - | 8 | 1413 | |||
BC021277.2 | - | 7 | 1307 | 4857987 |