Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284406.1 | NP_001271335.1 | 12 | 1398 | ||
NM_001284407.1 | NP_001271336.1 | 11 | 1220 | |||
NM_001284408.1 | NP_001271337.1 | 10 | 1082 | |||
NM_018306.3 | NP_060776.2 | 12 | 1310 | |||
GenBank mRNA | AK001898.1 | - | 11 | 1152 | ||
AK092470.1 | - | 12 | 1310 |