Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018323.3 | NP_060793.2 | 10 | 1461 | ||
XM_005248174.1 | XP_005248231.1 | 10 | 1444 | |||
XM_005248175.4 | XP_005248232.1 | 10 | 1239 | |||
GenBank mRNA | AF411320.1 | - | 10 | 1328 | ||
AK001967.1 | - | 10 | 1449 | |||
AK023186.1 | - | 3 | 242 | |||
AK128622.1 | - | 11 | 1366 | |||
AK296302.1 | - | 10 | 1151 | |||
AY065990.1 | - | 10 | 1439 | |||
AY091514.1 | - | 10 | 1448 | |||
BC051749.1 | - | 10 | 1461 | 6050442 |