Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164496.1 | NP_001157968.1 | 4 | 420 | ||
NM_018338.3 | NP_060808.2 | 4 | 420 | |||
XM_006713696.2 | XP_006713759.1 | 5 | 1010 | |||
XM_011512977.2 | XP_011511279.1 | 5 | 1010 | |||
XM_011512980.2 | XP_011511282.1 | 5 | 1010 | |||
XM_011512981.1 | XP_011511283.1 | 5 | 1010 | |||
XM_011512982.2 | XP_011511284.1 | 5 | 1010 | |||
XM_011512983.2 | XP_011511285.1 | 5 | 1010 | |||
XM_011512984.2 | XP_011511286.1 | 5 | 1010 | |||
XM_017006842.1 | XP_016862331.1 | 5 | 1010 | |||
XM_017006843.1 | XP_016862332.1 | 5 | 1010 | |||
XM_017006844.1 | XP_016862333.1 | 5 | 1010 | |||
XM_017006845.1 | XP_016862334.1 | 5 | 1010 | |||
XM_017006847.1 | XP_016862336.1 | 1 | 21 | |||
XM_017006848.1 | XP_016862337.1 | 5 | 1010 | |||
XM_017006849.1 | XP_016862338.1 | 5 | 1010 | |||
GenBank mRNA | AK056472.1 | - | 4 | 399 | ||
BC101582.1 | - | 4 | 378 | 8069088 | ||
BC113553.1 | - | 4 | 379 | 8322605 | ||
DB050626.1 | - | 4 | 420 |