Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018447.3 | NP_060917.1 | 4 | 752 | ||
XM_005265321.3 | XP_005265378.1 | 4 | 620 | |||
GenBank mRNA | AF157321.1 | - | 3 | 464 | ||
AK172843.1 | - | 3 | 460 | |||
AK222953.1 | - | 3 | 462 | |||
AK312008.1 | - | 3 | 459 | |||
BC022807.1 | - | 3 | 447 | 4897577 |