Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282857.1 | NP_001269786.1 | 14 | 1665 | ||
NM_019001.4 | NP_061874.3 | 14 | 1665 | |||
XM_006713673.2 | XP_006713736.1 | 14 | 1665 | |||
XM_011512919.2 | XP_011511221.1 | 14 | 1665 | |||
XM_011512920.2 | XP_011511222.1 | 14 | 1665 | |||
XM_011512922.2 | XP_011511224.1 | 11 | 1296 | |||
XM_017006640.1 | XP_016862129.1 | 14 | 1665 | |||
XM_017006641.1 | XP_016862130.1 | 14 | 1665 | |||
XM_017006642.1 | XP_016862131.1 | 14 | 1665 | |||
GenBank mRNA | AK123897.1 | - | 11 | 1138 | ||
AY137776.1 | - | 14 | 1547 | |||
BC078146.1 | - | 14 | 1577 | |||
BX640905.1 | - | 14 | 1614 | |||
CR749518.1 | - | 14 | 1595 |