Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019002.3 | NP_061875.2 | 2, 3 | 492 | ||
XM_017004376.1 | XP_016859865.1 | 2, 3 | 485 | |||
XM_017004377.1 | XP_016859866.1 | 2, 3 | 485 | |||
GenBank mRNA | AJ242682.2 | - | 2, 3 | 497 | ||
AK025155.1 | - | 1, 2 | 25 | |||
BC033075.1 | - | 2, 3 | 464 | |||
BC040001.1 | - | 2, 3 | 424 | 5556133 |