Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 6 | 1653 | ||
NM_019008.5 | NP_061881.2 | 6 | 1703 | |||
NR_130789.1 | 6 | 1617 | ||||
NR_130790.1 | 7 | 1767 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 1771 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 1891 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 1975 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 1869 | |||
GenBank mRNA | AK000239.1 | - | 3 | 874 | ||
AL136768.1 | - | 6 | 1653 | |||
AL365515.1 | - | 6 | 1580 | |||
AL834205.1 | - | 6 | 1545 | |||
BC002587.2 | - | 2 | 845 | |||
BC008327.1 | - | 6 | 1557 | 3502711 |