Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 7 | 3767 | ||
NM_019008.5 | NP_061881.2 | 6 | 4822 | |||
NR_130789.1 | 6 | 4736 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 4890 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 5010 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 5094 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 4988 | |||
GenBank mRNA | AF086364.1 | - | 1 | 511 |