Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019022.3 | NP_061895.3 | 16 | 1353 | ||
XM_005266714.2 | XP_005266771.1 | 15 | 1298 | |||
XM_011526033.2 | XP_011524335.1 | 14 | 1216 | |||
GenBank mRNA | AB058733.1 | - | 16 | 1315 | ||
AK000800.1 | - | 10 | 793 | |||
AK122715.1 | - | 16 | 1345 | |||
AK310389.1 | - | 16 | 1535 | |||
AL832341.1 | - | 16 | 1301 | |||
AL832356.1 | - | 16 | 1301 | |||
BC007690.2 | - | 15 | 1144 | |||
BC018981.2 | - | 15 | 1144 | |||
BC093792.1 | - | 16 | 1313 | 7939637 | ||
BC093794.1 | - | 16 | 1314 | 7939639 | ||
BC107422.1 | - | 16 | 1349 | |||
BC143400.1 | - | 15 | 1268 | |||
BX647846.1 | - | 16 | 1362 |