Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 9, 10 | 929 | ||
XM_006712600.2 | XP_006712663.1 | 9, 10 | 929 | |||
XM_011511359.1 | XP_011509661.1 | 9, 10 | 929 | |||
XM_011511361.1 | XP_011509663.1 | 8, 9 | 683 | |||
XM_011511362.1 | XP_011509664.1 | 8, 9 | 797 | |||
XM_011511363.2 | XP_011509665.1 | 9, 10 | 929 | |||
GenBank mRNA | AB209993.1 | - | 9, 10 | 848 | ||
AK025611.1 | - | 8, 9 | 697 | |||
AK057926.1 | - | 9, 10 | 790 | |||
AK290722.1 | - | 9, 10 | 836 | |||
AK309040.1 | - | 9, 10 | 836 | |||
BC028609.1 | - | 9, 10 | 902 | 4826022 | ||
BC093018.1 | - | 9, 10 | 819 | 30337330 |