Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 23 | 1979 | ||
XM_011511359.1 | XP_011509661.1 | 23 | 1976 | |||
XM_011511361.1 | XP_011509663.1 | 22 | 1733 | |||
XM_011511362.1 | XP_011509664.1 | 22 | 1847 | |||
GenBank mRNA | AB209993.1 | - | 23 | 1898 | ||
AK001858.1 | - | 11 | 773 | |||
AK025611.1 | - | 22 | 1747 | |||
AK290722.1 | - | 23 | 1886 | |||
BC028609.1 | - | 23 | 1952 | 4826022 | ||
BC093018.1 | - | 23 | 1869 | 30337330 |