Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256826.1 | NP_001243755.1 | 2, 3 | 479 | ||
NM_019088.3 | NP_061961.2 | 3, 4 | 509 | |||
GenBank mRNA | AJ401156.1 | - | 3, 4 | 334 | ||
AK098423.1 | - | 2, 3 | 276 | |||
BC000017.2 | - | 3, 4 | 353 | 3504227 | ||
BC013402.1 | - | 3, 4 | 425 | 2960015 |