Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259057.2 | XP_005259114.1 | 6, 7 | 1041 | ||
XM_011527113.1 | XP_011525415.1 | 6, 7 | 796 | |||
XM_011527114.1 | XP_011525416.1 | 6, 7 | 734 | |||
XM_011527115.1 | XP_011525417.1 | 6, 7 | 802 | |||
XM_011527116.1 | XP_011525418.1 | 6, 7 | 928 | |||
GenBank mRNA | AK022948.1 | - | 6, 7 | 1013 | ||
AK309830.1 | - | 6, 7 | 918 | |||
AL136606.1 | - | 6, 7 | 967 | |||
BC008869.2 | - | 6, 7 | 953 | 4131601 |