Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPCPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019593.3 | NP_062539.1 | 8 | 891 | ||
XM_005260758.4 | XP_005260815.1 | 9 | 1042 | |||
XM_005260761.1 | XP_005260818.1 | 7 | 815 | |||
XM_006723596.3 | XP_006723659.1 | 9 | 1815 | |||
GenBank mRNA | AB037855.1 | - | 8 | 882 | ||
AK308111.1 | - | 8 | 889 | |||
AL833069.1 | - | 3 | 2087 | |||
BC027588.2 | - | 8 | 890 | 4827346 |