Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020143.2 | NP_064528.1 | 5 | 603 | ||
GenBank mRNA | AF164799.1 | - | 5 | 561 | ||
AF349314.1 | - | 5 | 583 | |||
AK223118.1 | - | 5 | 531 | |||
AK225641.1 | - | 5 | 527 | |||
AK291715.1 | - | 5 | 591 | |||
AK313204.1 | - | 5 | 551 | |||
BC008304.1 | - | 5 | 603 | 3344225 | ||
BC018152.1 | - | 5 | 398 | |||
L48692.1 | - | 5 | 538 |