Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020202.4 | NP_064587.1 | 6 | 562 | ||
GenBank mRNA | AF260334.1 | - | 6 | 561 | ||
AF284574.1 | - | 6 | 509 | |||
AK225979.1 | - | 5 | 1040 | |||
AK295835.1 | - | 5 | 429 | |||
AK313704.1 | - | 6 | 504 | |||
BC020620.1 | - | 6 | 519 | 4775062 | ||
BC107890.1 | - | 6 | 550 | 6648910 | ||
FJ224326.1 | - | 6 | 509 |