Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127893.1 | NP_001121365.1 | 2 | 708 | ||
NM_020219.3 | NP_064604.2 | 2 | 708 | |||
GenBank mRNA | AK027157.1 | - | 1 | 291 | ||
AK291588.1 | - | 2 | 409 | |||
AY358394.1 | - | 2 | 438 | |||
BC083499.1 | - | 2 | 708 | 30390224 | ||
KJ851726.1 | - | 2 | 228 | |||
KJ851727.1 | - | 2 | 228 | |||
KJ851728.1 | - | 2 | 228 | |||
KJ851729.1 | - | 2 | 228 | |||
KJ851730.1 | - | 2 | 228 | |||
KJ851731.1 | - | 2 | 228 | |||
KJ851732.1 | - | 2 | 228 | |||
KJ851733.1 | - | 2 | 228 | |||
KJ851734.1 | - | 2 | 228 | |||
KJ851735.1 | - | 2 | 228 | |||
KJ851736.1 | - | 2 | 228 | |||
KJ851737.1 | - | 2 | 228 | |||
KJ851738.1 | - | 2 | 228 | |||
KJ851739.1 | - | 2 | 228 | |||
KJ851740.1 | - | 2 | 228 | |||
KJ851741.1 | - | 2 | 228 | |||
KJ851742.1 | - | 2 | 228 | |||
KJ851743.1 | - | 2 | 228 | |||
KJ851744.1 | - | 2 | 228 | |||
KJ851745.1 | - | 2 | 228 | |||
KJ851746.1 | - | 2 | 228 | |||
KJ851747.1 | - | 2 | 228 | |||
KJ851748.1 | - | 2 | 228 | |||
KJ851749.1 | - | 2 | 228 | |||
KJ851750.1 | - | 2 | 228 | |||
KJ851751.1 | - | 2 | 228 | |||
KJ851752.1 | - | 2 | 228 | |||
KJ851753.1 | - | 2 | 228 | |||
KJ851754.1 | - | 2 | 228 | |||
KJ851755.1 | - | 2 | 228 | |||
KJ851756.1 | - | 2 | 228 | |||
KJ851757.1 | - | 2 | 228 | |||
KJ851758.1 | - | 2 | 228 | |||
KJ851759.1 | - | 2 | 228 | |||
KJ851760.1 | - | 2 | 228 | |||
KJ851761.1 | - | 2 | 228 | |||
KJ851762.1 | - | 2 | 228 | |||
KJ851763.1 | - | 2 | 228 | |||
KJ851764.1 | - | 2 | 228 | |||
KJ851765.1 | - | 2 | 228 | |||
KJ851766.1 | - | 2 | 228 | |||
KJ851767.1 | - | 2 | 228 | |||
KJ851768.1 | - | 2 | 228 | |||
KJ851769.1 | - | 2 | 228 | |||
KJ851770.1 | - | 2 | 228 | |||
KJ851771.1 | - | 2 | 228 | |||
KJ851772.1 | - | 2 | 228 | |||
KJ851773.1 | - | 2 | 228 | |||
KJ851774.1 | - | 2 | 228 | |||
KJ851775.1 | - | 2 | 228 | |||
KJ851776.1 | - | 2 | 228 | |||
KJ851777.1 | - | 2 | 228 | |||
KJ851778.1 | - | 2 | 228 | |||
KJ851779.1 | - | 2 | 228 | |||
KJ851780.1 | - | 2 | 228 | |||
KJ851781.1 | - | 2 | 228 | |||
KJ851782.1 | - | 2 | 228 | |||
KJ851783.1 | - | 2 | 228 | |||
KJ851784.1 | - | 2 | 228 | |||
KJ851785.1 | - | 2 | 228 | |||
KJ851786.1 | - | 2 | 228 | |||
KJ851787.1 | - | 2 | 228 | |||
KJ851788.1 | - | 2 | 228 | |||
KJ851789.1 | - | 2 | 228 | |||
KJ851790.1 | - | 2 | 228 | |||
KJ851791.1 | - | 2 | 228 | |||
KJ851792.1 | - | 2 | 228 | |||
KJ851793.1 | - | 2 | 228 | |||
KJ851794.1 | - | 2 | 228 | |||
KJ851795.1 | - | 2 | 228 | |||
KJ851796.1 | - | 2 | 228 | |||
KJ851797.1 | - | 2 | 228 |